Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene Glycol Antibody (RM105), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB293884 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB293884 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB293884 Supplier Novus Biologicals Supplier No. NBP326041AF750
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Polyethylene Glycol Monoclonal antibody specifically detects Polyethylene Glycol in All samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Polyethylene Glycol
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM105
Conjugate Alexa Fluor 750
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias 1,2-Dihydroxyethane, 1,2-ethanediol, 107-21-1, 2-hydroxyethanol, Ethane-1,2-diol, glycol, Glycol alcohol, monoethylene glycol
Host Species Rabbit
Immunogen KLH conjugated Polyethylene Glycol (PEG) with terminal methoxy group
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species All species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.