Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Polygodial
Click to view available options
Quantity:
10 mg
50 mg
Description
TRPA1 channel activator (EC50 = 400 nM) that is selective against TRPV1, TRPV2, TRPV3, TRPV4 and TRPM8 channels. Acts as an analgesic via desensitization of sensory neurons. Also exhibits antifungal activity via inhibition of mitochondrial ATPase.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 234.33 |
Purity | >97% |
Molecular Weight (g/mol) | 234.34 |
Chemical Name or Material | Polygodial |
Safety and Handling
Recommended Storage | Store at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction