Learn More
Thermo Scientific™ Polymer Tubing Cutter
GSA_VA
Catalog No. p-6817046
Click to view available options
Type:
Tubing Cutter
Produce flat, 90°, burr-free ends of polymer tubing for HPLC applications using Thermo Scientific™ Polymer Tubing Cutter.
Produce flat, 90°, burr-free ends of polymer tubing for HPLC applications using the Thermo Scientific™ Polymer Tubing Cutter. It is compatible with rigid polymeric tubing and comes with guide holes for 1/16 in. and 1/8 in. tubing.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Type Tubing Cutter
Description 90° Burr-Free End
Quantity 1 Each
For Use With (Equipment) Tubing
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.