Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polymyxin B Antibody (45), Novus Biologicals™
SDP

Catalog No. NB119841 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119841 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119841 Supplier Novus Biologicals Supplier No. NBP311216
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Polymyxin B Monoclonal specifically detects Polymyxin B in Human samples. It is validated for Western Blot, Immunoassay.

Specifications

Antigen Polymyxin B
Applications Western Blot, Immunoassay
Classification Monoclonal
Clone 45
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Immunoassay
Formulation Culture Medium
Gene Alias Aerosporin, Poly-Rx
Host Species Mouse
Immunogen The monoclonal antibody 45 reacts with Polymyxin B.
Purification Method Unpurified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.