Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polynuclear Aromatic Hydrocarbons Antibody (BAP-13), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB50479A350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB50479A350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB50479A350 Supplier Novus Biologicals Supplier No. NB500479AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Polynuclear Aromatic Hydrocarbons Monoclonal specifically detects Polynuclear Aromatic Hydrocarbons in Non-species specific samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Polynuclear Aromatic Hydrocarbons
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone BAP-13
Conjugate Alexa Fluor 350
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias PAHs
Host Species Mouse
Immunogen Benzo[a]pyrenyl-1-butyric acid conjugated to BSA.
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity The antibody BAP-13 is specific for Polynuclear Aromatic Hydrocarbons. The antibody BAP-13 inhibits DNA adduct formation (this ability was investigated in rat liver microsomes spiked with calf thymus DNA and 7,8-diol-B[a]P).
Target Species Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.