Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polynuclear Aromatic Hydrocarbons Antibody (BAP-13), Biotin, Novus Biologicals™
SDP

Catalog No. NB359225 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359225 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359225 Supplier Novus Biologicals Supplier No. NB500479B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Polynuclear Aromatic Hydrocarbons Monoclonal antibody specifically detects Polynuclear Aromatic Hydrocarbons in Nonspecies specific samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Polynuclear Aromatic Hydrocarbons
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone BAP-13
Conjugate Biotin
Dilution ELISA, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias PAHs
Host Species Mouse
Immunogen Benzo[a]pyrenyl-1-butyric acid conjugated to BSA.
Purification Method Multi-step
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.