Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyoma Virus Early Antigen Antibody (PyC), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB102793AFT Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB102793AFT 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB102793AFT Supplier Novus Biologicals Supplier No. NB1002793AF700
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Polyoma Virus Early Antigen Monoclonal specifically detects Polyoma Virus Early Antigen in Virus samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot.

Specifications

Antigen Polyoma Virus Early Antigen
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone PyC
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Large T antigen, Middle T antigen, Polyoma virus large T antigen, Polyoma virus middle T antigen, Polyoma virus small T antigen, Polyomavirus early antigen, Polyomavirus large T antigen, Polyomavirus middle t antigen, Polyomavirus small T antigen, Small T antigen
Host Species Rat
Immunogen Polyoma virus-transformed Wistar rat fibroblast cellline REWA5/T1A1.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity Allows the detection and isolation of all three viral T antigens.
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.