Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyoma Virus, Medium T Antigen Antibody (PyMT), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB1002749FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002749FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002749FR Supplier Novus Biologicals Supplier No. NB1002749FR
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Polyoma Virus, Medium T Antigen Monoclonal specifically detects Polyoma Virus, Medium T Antigen in Virus samples. It is validated for Dual RNAScope ISH-IHC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Polyoma Virus, Medium T Antigen
Applications Dual RNAScope ISH-IHC, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone PyMT
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Alias Middle t antigen, MPyVgp2, Polyoma virus middle T antigen, Polyomavirus medium t antigen, Polyomavirus middle t antigen
Host Species Rat
Immunogen Polyoma Virus-transformed Wistar rat fibroblast cell line Py REWA5/T1A1.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Polyoma Virus.
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.