Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyoma Virus, Medium T Antigen Antibody (PyMT), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB339742 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339742 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339742 Supplier Novus Biologicals Supplier No. NB1002749MFV450
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Polyoma Virus, Medium T Antigen Monoclonal antibody specifically detects Polyoma Virus, Medium T Antigen in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Dual RNAscope ISH-IHC

Specifications

Antigen Polyoma Virus, Medium T Antigen
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PyMT
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Dual RNAscope ISH-IHC
Formulation 50mM Sodium Borate
Gene Alias Middle t antigen, MPyVgp2, Polyoma virus middle T antigen, Polyomavirus medium t antigen, Polyomavirus middle t antigen
Host Species Rat
Immunogen Polyoma Virus-transformed Wistar rat fibroblast cell line Py REWA5/T1A1.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.