Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polypeptide GalNac Transferase 10/GALNT10 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP177058 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177058 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177058 Supplier Novus Biologicals Supplier No. NBP177058
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Polypeptide GalNac Transferase 10/GALNT10 Polyclonal specifically detects Polypeptide GalNac Transferase 10/GALNT10 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Polypeptide GalNac Transferase 10/GALNT10
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_938080
Gene Alias DKFZp586H0623, EC 2.4.1.41, FLJ00205, FLJ11715, GalNAc transferase 10, GalNAc-T10, GALNACT10, Polypeptide GalNAc transferase 10, polypeptide N-acetylgalactosaminyltransferase 10, PPGALNACT10, pp-GaNTase 10, PPGANTASE10, Protein-UDP acetylgalactosaminyltransferase 10, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 10, UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 10 (GalNAc-T10)
Gene Symbols GALNT10
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human GALNT10. The immunogen is located within amino acids 530 - 580 of GALNT10. Amino Acid Squence: KGNQLWKYRKDKTLYHPV
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55568
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.