Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyuridylic Acid, Potassium Salt, MP Biomedicals™

Catalog No. ICN10270710 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10270710 10 mg
ICN10270705 5 mg
ICN10270725 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10270710 Supplier MP Biomedicals, Inc Supplier No. 0210270710

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Polyuridylic acid is an RNA analog, a homopolymer of uridine

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28086-43-3
Molecular Formula C9H11N2O9P-2
Molecular Weight (g/mol) 322.166
MDL Number MFCD00148052
InChI Key DJJCXFVJDGTHFX-UHFFFAOYSA-L
Synonym disodium 5-2,4-dioxopyrimidin-1-yl-3,4-dihydroxy-oxolan-2-yl methoxyphosphonic acid, 5-2,4-dioxopyrimidin-1-yl-3,4-dihydroxyoxolan-2-yl methyl phosphate
PubChem CID 4014956
IUPAC Name [5-(2,4-dioxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES C1=CN(C(=O)NC1=O)C2C(C(C(O2)COP(=O)([O-])[O-])O)O

Specifications

Color White
Quantity 10 mg
Formula Weight >100,000
Physical Form Solid
Chemical Name or Material Polyuridylic acid potassium salt
Recommended Storage Store at -0°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.