Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GVS Polyvinylidene Fluoride Sheet
Click to view available options
Pore Size:
0.22 μm
0.45 μm
Dimensions (LxW):
100 x 100 mm
150 x 150 mm
200 x 200 mm
Description
Properties
- This membrane also has broad chemical compatibility, which is important when used with common stains such as Amido Black, Colloidal Gold, Coomassie Blue, India Ink and Ponceau-S.
- This Polyvinylidene fluoride membrane will not degrade, distort or shrink when a high concentration of methanol is used for destaining. Its exceptional strength, high binding capacity and chemical compatibility make GVS Polyvinylidene fluoride membrane ideal for use in Western blots, immunoblotting, and solid phase assays and plaque lifts.
Characteristics
- Superior strength can withstand aggressive handling or be used with automated equipment without breaking or tearing
- Low extractables ensures tests will be clean with consistent results
- Exceptional sensitivity detects low-level components
- Hydrophobic for high protein binding
- Lot-to-lot consistency quality checks ensure consistent binding for dependable results every time
- BSA protein binding capacity 125μg/cm²
Applications
- Western blots
- Immunoblotting
- Solid phase assays
- Amino acid or protein analyses
Specifications
Specifications
Length (Metric) | 200 mm |
Width (Metric) | 200 mm |
Format | Sheet |
Material | PVDF |
Pore Size | 0.22 μm |
Dimensions (LxW) | 200 x 200 mm |
Thickness | 140 to 250 μm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction