Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POMGNT1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | POMGNT1 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
POMGNT1 Polyclonal specifically detects POMGNT1 in Mouse samples. It is validated for Western Blot.Specifications
| POMGNT1 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| DKFZp761B182, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.101, FLJ20277, GnT I.2, MDDGA3, MDDGB3, MDDGC3, MEB, MGAT1.2GNTI.2, POMGnT1, protein O-linked mannose beta1,2-N-acetylglucosaminyltransferase, protein O-linked-mannose beta-1,2-N-acetylglucosaminyltransferase 1, UDP-GlcNAc:alpha-D-mannoside beta-1,2-N-acetylglucosaminyltransferase I.2 | |
| POMGNT1 | |
| IgG |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Q91X88 | |
| 55624 | |
| Synthetic peptides corresponding to the N terminal of Pomgnt1. Immunizing peptide sequence LLVTVIVNIKLILDTRRAISEANEDPEPEQDYDEALGRLESPRRRGSSPR. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title