Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POMK/SGK196 Antibody (OTI2B10), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB272275A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB272275A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB272275A48 Supplier Novus Biologicals Supplier No. NBP272275AF488

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

POMK/SGK196 Monoclonal specifically detects POMK/SGK196 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen POMK/SGK196
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2B10
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ23356, MGC126597, protein kinase-like protein SgK196, Sugen kinase 196
Gene Symbols POMK
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 141-350 of human FLJ23356 (NP_115613) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 84197.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.