Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PON1 Antibody (4G8D3), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB005172 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB005172 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB005172 Supplier Novus Biologicals Supplier No. NBP223610JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PON1 Monoclonal specifically detects PON1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.

Specifications

Antigen PON1
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 4G8D3
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias A-esterase 1, Aromatic esterase 1, arylesterase B-type, EC 3.1.1.2, EC 3.1.1.81, EC 3.1.8.1, ESAesterase A, K-45, MVCD5, paraoxonase 1, PON 1, PONparaoxonase B-type, serum aryldiakylphosphatase, Serum aryldialkylphosphatase 1, serum paraoxonase/arylesterase 1
Gene Symbols PON1
Host Species Mouse
Immunogen Partial recombinant human PON1 (between residues 1-200) expressed in E. coli [Uniprot P27169]
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cancer, Lipid and Metabolism, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.