Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PON1 Antibody (4G8D3), Novus Biologicals™
SDP

Catalog No. NBP223610 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223610 0.1 mL
NBP223610SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP223610 Supplier Novus Biologicals Supplier No. NBP223610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PON1 Monoclonal specifically detects PON1 in Human samples. It is validated for Flow Cytometry, ICC/IF (Negative), Immunohistochemistry, Western Blot.

Specifications

Antigen PON1
Applications Flow Cytometry, ICC/IF (Negative), Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4G8D3
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, Immunohistochemistry 1:200 - 1:1000, ICC/IF (Negative)
Gene Accession No. P27169
Gene Alias A-esterase 1, Aromatic esterase 1, arylesterase B-type, EC 3.1.1.2, EC 3.1.1.81, EC 3.1.8.1, ESAesterase A, K-45, MVCD5, paraoxonase 1, PON 1, PONparaoxonase B-type, serum aryldiakylphosphatase, Serum aryldialkylphosphatase 1, serum paraoxonase/arylesterase 1
Gene Symbols PON1
Host Species Mouse
Immunogen Partial recombinant human PON1 (between residues 1-200) expressed in E. coli [Uniprot P27169]
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cancer, Lipid and Metabolism, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 5444
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Precipitation
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.