Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PON1 Antibody (OTI2D4), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP273526G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273526G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273526G Supplier Novus Biologicals Supplier No. NBP273526G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PON1 Monoclonal specifically detects PON1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PON1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2D4
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias A-esterase 1, Aromatic esterase 1, arylesterase B-type, EC 3.1.1.2, EC 3.1.1.81, EC 3.1.8.1, ESAesterase A, K-45, MVCD5, paraoxonase 1, PON 1, PONparaoxonase B-type, serum aryldiakylphosphatase, Serum aryldialkylphosphatase 1, serum paraoxonase/arylesterase 1
Gene Symbols PON1
Host Species Mouse
Immunogen Full length recombinant protein of human PON1 (NP_000437) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Alzheimers Research, Cancer, Lipid and Metabolism, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 5444.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.