Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POR/Cytochrome P450 Reductase Antibody, Novus Biologicals™
SDP

Catalog No. NBP152083 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152083 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152083 Supplier Novus Biologicals Supplier No. NBP152083
Add to Cart
Add to Cart

Goat Polyclonal Antibody

POR/Cytochrome P450 Reductase Polyclonal specifically detects POR/Cytochrome P450 Reductase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen POR/Cytochrome P450 Reductase
Applications Western Blot, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.1 - 0.3 ug/mL, Immunohistochemistry-Paraffin 3.75 ug/mL, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_000932.3
Gene Alias CPR, CYPORFLJ26468, DKFZp686G04235, EC 1.6.2.4, NADPH--cytochrome P450 reductase, NADPH-dependent cytochrome P450 reductase, P450 (cytochrome) oxidoreductase, P450R
Gene Symbols POR
Host Species Goat
Immunogen internal region
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5447
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Rabbit
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.