Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POR/Cytochrome P450 Reductase Antibody (OTI3F10), PE, Novus Biologicals™
SDP

Catalog No. NBP273539PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273539PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273539PE Supplier Novus Biologicals Supplier No. NBP273539PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

POR/Cytochrome P450 Reductase Monoclonal specifically detects POR/Cytochrome P450 Reductase in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen POR/Cytochrome P450 Reductase
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3F10
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CPR, CYPORFLJ26468, DKFZp686G04235, EC 1.6.2.4, NADPH--cytochrome P450 reductase, NADPH-dependent cytochrome P450 reductase, P450 (cytochrome) oxidoreductase, P450R
Gene Symbols POR
Host Species Mouse
Immunogen Full length recombinant protein of human POR/Cytochrome P450 Reductase (NP_000932) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Breast Cancer, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5447.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.