Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Porcine IL-8/CXCL8 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
$200.50 - $1973.00
Specifications
Antigen | CXCL8/IL-8 |
---|---|
Clone | 105105 |
Dilution | Western Blot 1 ug/mL, Neutralization 1-6 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Western Blot, Neutralization, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB5351SP
![]() |
R&D Systems
MAB5351SP |
25 μg |
Each for $200.50
|
|
|||||
MAB5351100
![]() |
R&D Systems
MAB5351100 |
100 μg |
Each for $538.00
|
|
|||||
MAB5351500
![]() |
R&D Systems
MAB5351500 |
500 μg |
Each for $1,973.00
|
|
|||||
Description
CXCL8/IL-8 Monoclonal specifically detects CXCL8/IL-8 in Porcine samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
CXCL8/IL-8 | |
Western Blot 1 ug/mL, Neutralization 1-6 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Mouse | |
Pig | |
CAA43461 | |
3576 | |
E. coli-derived recombinant porcine IL-8/CXCL8 Ala26-Gln104 Accession # CAA43461 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects porcine IL-8/CXCL8 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) CXCL1, 2, 3, 5, 6, 7, 8, 9, 10, 11, 12/SDF-1 alpha , 12/SDF-1 beta , rhCXCL13, recombinant mouse CXCL1, 2, 6, 9, 10, 12/SDF-1 alpha , rmCXCL13, recombinant rat (rr) CXCL1, rrCXCL3/CINC2 alpha , or rr3/CINC-2 beta is observed. |
105105 | |
Western Blot, Neutralization, ELISA | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1 | |
IL8 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title