Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Porimin Antibody (297617), PerCP, Novus Biologicals™
SDP

Catalog No. NB363505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363505 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363505 Supplier Novus Biologicals Supplier No. FAB3010C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Porimin Monoclonal specifically detects Porimin in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen Porimin
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 297617
Conjugate PerCP
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias KCT-3, KCT3keratinocytes associated transmembrane protein 3, Keratinocytes-associated transmembrane protein 3, PORIMINporimin, pro oncosis receptor inducing membrane injury, Pro-oncosis receptor inducing membrane injury, serine/threonine-rich receptor, transmembrane protein 123PORMIN
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Porimin, His27-Ser149, Accession # AAG21694
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 114908
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.