Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Porimin Antibody, Novus Biologicals™
SDP

Catalog No. NBP227172 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227172 0.1 mg
NBP227172SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227172 Supplier Novus Biologicals Supplier No. NBP227172
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Porimin Polyclonal specifically detects Porimin in Rat samples. It is validated for Western Blot.

Specifications

Antigen Porimin
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 5-8ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q91Z22
Gene Alias KCT-3, KCT3keratinocytes associated transmembrane protein 3, Keratinocytes-associated transmembrane protein 3, PORIMINporimin, pro oncosis receptor inducing membrane injury, Pro-oncosis receptor inducing membrane injury, serine/threonine-rich receptor, transmembrane protein 123PORMIN
Gene Symbols TMEM123
Host Species Rabbit
Immunogen 179 SRRGIRYRSIDEHDAII 195
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114908
Test Specificity The amino acid sequence used as immunogen is 100% homologous in rat and mouse, 94% in pig, new world monkey, human, cow, dog, orangutan, rhesus monkey, platypus and white-cheeked gibbon, and 88% homologous in horse, wild turkey, and chicken.
Target Species Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.