Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Porimin Antibody, Novus Biologicals™
SDP

Catalog No. NB10056371 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056371 0.1 mg
NB10056371T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056371 Supplier Novus Biologicals Supplier No. NB10056371
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

Porimin Polyclonal specifically detects Porimin in Bovine, Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Porimin
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500. Use reported in scientific literature (PMID 16814713)
Formulation PBS containing 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q8N131
Gene Alias KCT-3, KCT3keratinocytes associated transmembrane protein 3, Keratinocytes-associated transmembrane protein 3, PORIMINporimin, pro oncosis receptor inducing membrane injury, Pro-oncosis receptor inducing membrane injury, serine/threonine-rich receptor, transmembrane protein 123PORMIN
Gene Symbols TMEM123
Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acids 173-188 SRRGIRYRTIDEHDAI of human Porimin; GenBank no. AAG2164.1.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 114908
Test Specificity Reacts with Human and bovine. This sequence is 100% conserved in cow, and 93% conserved in rat, mouse, and chicken.
Target Species Bovine, Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.