Description
- Large crystals in variety of shapes and colors
- Felsic plutonic rock ideal for lessons in mineral identification and bulk rock chemistry
- For comparing cooling rates between samples with differently-sized crystals
- Correlates well to Next Generation Science Standards Evidence Statements
- Large crystals are indicative of slow cooling rate, and make mineral identification simple
Specifications
Specifications
| Product Type | Igneous Rock |
| For Use With (Application) | Comparing cooling rates between samples with differently-sized crystals |
| Packaging | Clear labeled and barcoded bag |
| Quantity | 12/Pk. |
| Specimen Size | 2 to 3 cm |
| Specimen Type | Porphyritic Granite |