Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POT1 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB500176B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500176B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500176B Supplier Novus Biologicals Supplier No. NB500176B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

POT1 Polyclonal specifically detects POT1 in Human samples. It is validated for Western Blot.

Specifications

Antigen POT1
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q9NUX5
Gene Alias DKFZp586D211, hPot1, POT1 protection of telomeres 1 homolog, POT1-like telomere end-binding protein, protection of telomeres 1 homolog (S. pombe), protection of telomeres protein 1
Gene Symbols POT1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human POT1 protein sequence (between residues 200-300). [UniProt# Q9NUX5]
Molecular Weight of Antigen 71.4 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 25913
Test Specificity Human. Immunogen sequence has 95% homology to monkey, 82% homology to rat, and 72% homology to mouse.
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.