Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potash, Sulfurated (Potassium Polysulfide), Reagents

Catalog No. 01337128
Change view
Click to view available options
:
Quantity: 500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-337-128 Quantity: 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-128 Supplier Reagents Holdings Llc Supplier No. C2275200500A5
Only null left
Add to Cart
Add to Cart

Potash, Sulfurated (Potassium Polysulfide), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10294-66-3 , 37488-75-8
Molecular Formula K2O3S2
Molecular Weight (g/mol) 190.31
InChI Key FGRVOLIFQGXPCT-UHFFFAOYSA-L
Synonym Liver of Sulfur
IUPAC Name dipotassium 1-oxo-1λ⁶-disulfene-1,1-bis(olate)
SMILES [K+].[K+].[O-]S([O-])(=O)=S

Specifications

Color Yellow-Brown to Olive Green to Gray
Physical Form Solid
Quantity 500 g
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.8
Chemical Name or Material Potash, Sulfurated (Potassium Polysulfide)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.