Learn More
Potassium antimonate trihydrate, 94+%
Description
Potassium antimonyl trihydrate is used as a detecting agent for sodium. It is used as a mordant/ fixing agent in leather and textile dying industries. It is also used as a analytical reagent and flux additive for electroplating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Physical Form | Crystals or Powder |
| Quantity | 50 g |
| Assay Percent Range | >94% |
| UN Number | 1549 |
| Synonym | Potassium Pyroantimonate, Potassium Hexahydroxyantimonate |
| Solubility Information | Soluble in water and glycerine. Insoluble in ethanol. |
| Formula Weight | 262.90 (208.85 Anhydrous) |
| Appearance | White powder |
| Chemical Name or Material | Potassium Antimonate Trihydrate |
Safety and Handling
| Hazard Category | H302+H332 |
| Precautionary Statement | P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c |
| DOTInformation | DOT Class: 6.1, Packaging Group: III |
| EINECSNumber | 235-387-7 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.