Alfa Aesar™ Potassium bromide, Premium FTIR Grade

Manufacturer: Alfa Aesar™ 47111KD

Quantity 50 pieces

 View more versions of this product

Catalog No. AA47111KD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7758-02-3
    Linear Formula KBr
    Formula Weight 119.01
    MDL Number MFCD00011358
    Merck Index 14,7618
    Chemical Name or Material Potassium bromide
    Grade Premium FTIR Grade

    Specifications

    Formula Weight 119.01
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00011358
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Odor Odorless
    Physical Form Powder
    Refractive Index 1.559
    CAS 7758-02-3
    Merck Index 14,7618
    Boiling Point 1435°C
    Packaging 0.5g packets
    Linear Formula KBr
    Health Hazard 1 Warning
    Density 2.75g/mL
    TSCA TSCA
    Melting Point 730°C
    Quantity 50 pieces
    RTECS Number TS7650000
    Chemical Name or Material Potassium bromide
    Grade Premium FTIR Grade

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit