Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium bromide, Puratronic™, 99.999% (metals basis)

Catalog No. p-7048546
Click to view available options
Quantity:
10 g
50 g

CAS: 7758-02-3 | BrK | 119.00 g/mol

Potassium bromide is used in a variety of applications such as spectroscopy, infrared transmission, as a laboratory reagent, and for the removal of peripheral membrane proteins. It is also used in photosensitive chemicals, paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-02-3
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
SMILES [K+].[Br-]

Specifications

Boiling Point 1435°C
Melting Point 730°C
Color White
Physical Form Crystalline
Quantity 10 g
Linear Formula KBr
Merck Index 14,7618
Solubility Information Soluble in water. Slightly soluble in alcohol,glycerol and ether.
IUPAC Name potassium;bromide
Formula Weight 119.01
Percent Purity 99.999%
Grade Puratronic™
Odor Odorless
Refractive Index 1.559
Sensitivity Hygroscopic
Density 2.75 g/mL
Assay (metals basis)
Chemical Name or Material Potassium bromide
Show More Show Less
Hazard Category H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
EINECSNumber 231-830-3
RTECSNumber TS7650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.