Learn More
Potassium bromide, Spectroscopy Grade
Description
Manufacturing of photographic papers, process engraving and lithography, spectroscopy, as an analytical lab reagent. Potassium bromide is applied as a veterinary drug and antiepileptic medication for dogs. It is an utilitarian for the bromination of alkenes using cerium(IV) ammonium nitrate. It acts as an essential role with diacetoxy iodobenzene for the efficient
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1435°C |
| Melting Point | 730°C |
| Color | White |
| Physical Form | Random Cuttings, Chunks |
| Quantity | 1 lb. |
| Merck Index | 14,7618 |
| Solubility Information | Soluble in water and very slightly soluble in diethyl ether. |
| Formula Weight | 119.01 |
| Grade | Spectroscopy |
| Odor | Odorless |
| Show More |
Safety and Handling
| Hazard Category | H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c |
| EINECSNumber | 231-830-3 |
| RTECSNumber | TS7650000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.