Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Citrate Monohydrate, Reagents

Catalog No. 01336862
Change view
Click to view available options
:
Quantity: 50 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-336-862 Quantity: 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-862 Supplier Reagents Holdings Llc Supplier No. C328150050F4
Only null left
Add to Cart
Add to Cart

Potassium Citrate Monohydrate, Meets FCC specifications, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6100-05-6
Molecular Formula C6H7K3O8
Molecular Weight (g/mol) 324.41
InChI Key PJAHUDTUZRZBKM-UHFFFAOYSA-K
Synonym Tripotassium Citrate Monohydrate
IUPAC Name tripotassium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
SMILES O.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

Boiling Point 230°C
Color Light Yellow
pH 7.5 to 9.0
Physical Form Solid
Quantity 50 kg
Packaging Fiber Drum
Specific Gravity 1.98
Chemical Name or Material Potassium Citrate Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.