Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium cyanate, 97%

Catalog No. AA3335622
Click to view available options
Quantity:
100 g
500 g
2 kg

Catalog No. AA3335622

Supplier: Thermo Scientific Chemicals 03335622

Only null left
Add to Cart

CAS: 590-28-3 | CKNO | 81.12 g/mol

Potassium cyanate is used in gold mining, organic synthesis and electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-28-3
Molecular Formula CKNO
Molecular Weight (g/mol) 81.12
MDL Number MFCD00011411
InChI Key GKKCIDNWFBPDBW-UHFFFAOYSA-M
Synonym potassium cyanate, kaliumcyanat, kocn, aero cyanate, cyanic acid, potassium salt, alicyanate, bulpur, potassiumcyanate, weedanol cyanol, caswell no. 688
PubChem CID 11378442
ChEBI CHEBI:38904
SMILES [K+].[O-]C#N

Specifications

Melting Point 700°C to 900°C (decomposition)
Color White
Physical Form Crystalline
Quantity 100 g
Merck Index 14,7625
Solubility Information Soluble in water; Slightly soluble in alcohol
IUPAC Name potassium;cyanate
Formula Weight 81.12
Percent Purity 97%
Odor Odorless
Density 2.056 g/mL
Chemical Name or Material Potassium cyanate
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 209-676-3
RTECSNumber GS6825000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.