Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium cyanate, 97%
Click to view available options
Quantity:
100 g
500 g
2 kg
Description
Potassium cyanate is used in gold mining, organic synthesis and electroplating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 700°C to 900°C (decomposition) |
| Color | White |
| Physical Form | Crystalline |
| Quantity | 500 g |
| Merck Index | 14,7625 |
| Solubility Information | Soluble in water; Slightly soluble in alcohol |
| IUPAC Name | potassium;cyanate |
| Formula Weight | 81.12 |
| Percent Purity | 97% |
| Odor | Odorless |
| Show More |
Safety and Handling
| Hazard Category | H302 |
| Hazard Statement | GHS H Statement H302 Harmful if swallowed. |
| Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
| EINECSNumber | 209-676-3 |
| RTECSNumber | GS6825000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction