Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium cyclopropyltrifluoroborate, tech. 90%

Catalog No. aah6067303
Click to view available options
Quantity:
1 g
5 g

CAS: 1065010-87-8 | C3H5BF3K | 147.977 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1065010-87-8
Molecular Formula C3H5BF3K
Molecular Weight (g/mol) 147.977
MDL Number MFCD09265031
InChI Key CFMLURFHOSOXRC-UHFFFAOYSA-N
Synonym potassium cyclopropyltrifluoroborate, potassium cyclopropyltrifluoroboranuide, potassium cyclopropyltrifluororate, potassium cyclopropytrifluoroborate, potassium cycloproyltrifluoroborate, potassiumcyclopropyltrifluoroborate, potassium cyclopropyl-trifluoroborate, cyclopropyltrifluoroborate potassium salt, potassium;cyclopropyl trifluoro boranuide, potassium ion cyclopropyltrifluoroboranuide
PubChem CID 23697338
IUPAC Name potassium;cyclopropyl(trifluoro)boranuide
SMILES [B-](C1CC1)(F)(F)F.[K+]

Specifications

Melting Point 348°C to 350°C
Color White
Physical Form Powder
Quantity 1 g
Formula Weight 147.98
Percent Purity 90%
Grade Technical
Sensitivity Air Sensitive
Chemical Name or Material Potassium cyclopropyltrifluoroborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.