Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium ethyltrifluoroborate, 97%

Catalog No. p-4602067
Click to view available options
Quantity:
1 g
5 g

CAS: 44248-07-9 | C2H5BF3K | 135.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 44248-07-9
Molecular Formula C2H5BF3K
Molecular Weight (g/mol) 135.97
MDL Number MFCD04112713
InChI Key GIIPADVFWNGSKY-UHFFFAOYSA-N
Synonym potassium ethyltrifluoroborate, potassium ethyltrifluoroboranuide, potassium ethytrifluoroborate, amtb738, ethyltrifluoroborate potassium salt, potassium ethyl trifluoro borate 1-
PubChem CID 23668491
IUPAC Name potassium;ethyl(trifluoro)boranuide
SMILES [K+].CC[B-](F)(F)F

Specifications

Melting Point 80.0°C to 84.0°C
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 135.97
Percent Purity 97%
Chemical Name or Material Potassium ethyltrifluoroborate
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.