Learn More
Potassium hydrogen phosphate, dried, 98+%
Description
Potassium hydrogen phosphate is used as a fertilizer, food additive, buffering agent, in molecular biology, biochemistry and chromatography. It is useful to increase the partition coefficients of amino acids in aqueous two-phase systems. It plays an important role for increase in rice grain yield by spraying chemicals including ferric chloride, dipotassium hydrogen phosphate and salicylic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium hydrogen phosphate is used as a fertilizer, food additive, buffering agent, in molecular biology, biochemistry and chromatography. It is useful to increase the partition coefficients of amino acids in aqueous two-phase systems. It plays an important role for increase in rice grain yield by spraying chemicals including ferric chloride, dipotassium hydrogen phosphate and salicylic acid.
Solubility
Soluble in water. Slightly soluble in alcohol.
Notes
Hygroscopic chemical and incompatible with strong oxidizing agents and acids. Keep the material away from heat, sparks and flame and sources of ignition.
Specifications
Specifications
| Quantity | 250 g |
| Merck Index | 14,7658 |
| Solubility Information | Soluble in water. Slightly soluble in alcohol. |
| Formula Weight | 174.18 |
| Percent Purity | ≥98% |
| Sensitivity | Hygroscopic |
| Chemical Name or Material | Potassium hydrogen phosphate, Dried |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H319 Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement P280-P264-P305+P351+P338-P337+P313 Wear protective gloves/protective clothing/eye protection/face protection. Wash thoroughly after handling. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. |
| EINECSNumber | 231-834-5 |
| RTECSNumber | TC5580000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.