Learn More
Potassium hydrogen phthalate, 99%
Description
Potassium hydrogen phthalate is used as a primary standard for acid- base titrations as well as for calibrating pH meters and for the preparation of volumetric alkali solutions. It acts as buffer in pH determination. It finds application as a useful standard for total organic carbon testing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium hydrogen phthalate is used as a primary standard for acid- base titrations as well as for calibrating pH meters and for the preparation of volumetric alkali solutions. It acts as buffer in pH determination. It finds application as a useful standard for total organic carbon testing.
Solubility
Soluble in water. Slightly soluble in alcohol.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 297°C (decomposition) |
| Density | 1.636 |
| Odor | Odorless |
| Quantity | 500 g |
| Beilstein | 3637128 |
| Merck Index | 14,7612 |
| Solubility Information | Soluble in water. Slightly soluble in alcohol. |
| Formula Weight | 204.23 |
| Percent Purity | 99% |
| Chemical Name or Material | Potassium hydrogen phthalate |
Safety and Handling
| EINECSNumber | 212-889-4 |
| RTECSNumber | CZ4326000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.