Learn More
Potassium Hydroxide, 0.0100 N (N/100) in Methanol, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Color | Colorless |
| Assay Percent Range | 0.01000 ± 0.00005 N at 20°C |
| Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
| Solubility Information | Solubility in Water: Infinite |
| Quantity | 4 L |
| Grade | Laboratory |
| Odor | Characteristic alcohol |
| Packaging | Cubitainer |
| Solvent or Matrix | Nonaqueous Solution |
| Physical Form | Liquid |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.