Learn More
Potassium Hydroxide, 0.500 N in Denatured Ethanol (Reagent Alcohol), Reagents
Specifications
Specifications
| pH | >13 |
| Color | Colorless |
| Boiling Point | 78°C |
| Quantity | 20 L |
| Grade | Reagent |
| Packaging | HDPE Winpak |
| Specific Gravity | 0.8168 |
| Physical Form | Liquid |
| Chemical Name or Material | Potassium Hydroxide, 0.500 N in Denatured Ethanol (Reagent Alcohol) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.