Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 0.500 Normal in Methanol, Reagents

Catalog No. 01336440
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-336-440 4 L Poly Bottle
01-336-443 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-440 Supplier Reagents Holdings Llc Supplier No. C72850304A
Add to Cart
Add to Cart

Potassium Hydroxide, 0.500 Normal in Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-56-1 , 1310-58-3 , 1310-58-3
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Caustic Potash, Potash Lye
IUPAC Name methanol
SMILES CO

Specifications

pH >13
Color Colorless
Boiling Point 65°C
Quantity 4 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.8108
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide, 0.500 Normal in Methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.