Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 5% w/v in 3A Ethanol, Reagents

Catalog No. 01334324
Change view
Click to view available options
:
Quantity: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-334-324 Quantity: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-324 Supplier Reagents Holdings Llc Supplier No. CP1187001A
Only null left
Add to Cart
Add to Cart

Potassium Hydroxide, 5% w/v in 3A Ethanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 7732-18-5 , 1310-58-3
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym Caustic Potash, Potash Lye
IUPAC Name ethanol
SMILES CCO

Specifications

pH >13
Color Colorless to Orange
Boiling Point 78°C
Quantity 1 L
Percent Purity 5%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.85
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide, 5% w/v in 3A Ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.