Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium methanesulfonate, 99%

Catalog No. p-7050805
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 2386-56-3 | CH3KO3S | 134.19 g/mol

Used in pharmaceutical, chemical and biological research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2386-56-3
Molecular Formula CH3KO3S
Molecular Weight (g/mol) 134.19
MDL Number MFCD00070544
InChI Key XWIJIXWOZCRYEL-UHFFFAOYSA-M
Synonym potassium methanesulfonate, methanesulfonic acid, potassium salt, potassium mesylate, potassium methylsulfonate, potassium methyl sulfonate, potassium methanesulphonate, unii-s99ib3em17, methanesulfonic acid potassium salt, methanesulfonic acid,potassium salt 1:1, methanesulfonic acid, potassium salt 1:1
PubChem CID 23666501
IUPAC Name potassium;methanesulfonate
SMILES CS(=O)(=O)[O-].[K+]

Specifications

Color White
Odor Odorless
Linear Formula KCH3SO3
Quantity 25 g
Beilstein 3914899
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 134.19
Percent Purity 99%
Physical Form Powder or Lumps
Chemical Name or Material Potassium methanesulfonate
Show More Show Less
EINECSNumber 219-202-7
RTECSNumber PB2675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.