Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Stannate, Reagents

Catalog No. 01334422
Click to view available options
:
Quantity: 100 lb.

Catalog No. 01-334-422

Supplier: Reagents Holdings Llc CP105300100LB

Only null left
Add to Cart
Add to Cart
Request a Quote

Potassium Stannate, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 12142-33-5
Molecular Formula K2O3Sn
Molecular Weight (g/mol) 244.90
InChI Key IOUCSUBTZWXKTA-UHFFFAOYSA-N
IUPAC Name dipotassium oxostannanebis(olate)
SMILES [K+].[K+].[O-][Sn]([O-])=O

Specifications

Color White
Physical Form Solid
Quantity 100 lb.
Grade Reagent
Packaging Fiber Drum
Specific Gravity 3.197
Chemical Name or Material Potassium Stannate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.