Potassium tetrachloroplatinate(II), 99.9% (metals basis), Pt 46.0% min., Alfa Aesar™

Manufacturer: Alfa Aesar 1104806

Quantity 5g

 View more versions of this product

Catalog No. AA1104806

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 10025-99-7
    Physical Form Crystalline
    Assay Percent Range 99.9% (metals basis)
    Molecular Formula Cl4K2Pt
    Formula Weight 415.11
    MDL Number MFCD00011378
    Chemical Name or Material Potassium tetrachloroplatinate(II), Pt 46.0% min.

    Specifications

    Formula Weight 415.11
    Quantity 5g
    Color Red-brown
    DOT Information DOT Class: 6.1, Packaging Group: III
    EINECS Number 233-050-9
    Physical Form Crystalline
    Odor Odorless
    Assay Percent Range 99.9% (metals basis)
    Molecular Formula Cl4K2Pt
    Appearance Red-brown crystals
    MDL Number MFCD00011378
    CAS 10025-99-7
    Health Hazard 1 H301-H315-H317-H334-H318-H290: Toxic if swallowed. Causes skin irritation. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes serious eye damage. May be corrosive to metals.
    Chemical Name or Material Potassium tetrachloroplatinate(II), Pt 46.0% min.

    H301-H315-H317-H334-H318-H290: Toxic if swallowed. Causes skin irritation. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes serious eye damage. May be corrosive to metals.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit