Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Tripolyphosphate, Spectrum™ Chemical
SDP

Catalog No. 18605741 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

P1669, 13845-36-8, K5P3O10

Spectrum™ Chemical Potassium Tripolyphosphate

Chemical Identifiers

CAS 13845-36-8
Molecular Formula K5O10P3
Molecular Weight (g/mol) 448.40
InChI Key ATGAWOHQWWULNK-UHFFFAOYSA-I
IUPAC Name pentapotassium bis(phosphonatooxy)phosphinate
SMILES [K+].[K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Grade Reagent
Packaging Poly Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.