Learn More
Description
Specifications
Specifications
| Antigen | POTEA |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | A26A1, ANKRD26-like family A, member 1, CT104.3, POTE ankyrin domain family member A, POTE ankyrin domain family, member A, POTE-8ANKRD26-like family A member 1, POTE8cancer/testis antigen family 104, member 3, Prostate, ovary, testis-expressed protein on chromosome 8, protein expressed in prostate, ovary, testis, and placenta 8 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the middle region of Human POTEA |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
