Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POTEE Antibody, Novus Biologicals™
SDP

Catalog No. NBP254706 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP254706 100 μL
NB395293 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP254706 Supplier Novus Biologicals Supplier No. NBP254706
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

POTEE Polyclonal specifically detects POTEE in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen POTEE
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias A26C1, A26C1A, ANKRD26-Like Family C Member 1A, ANKRD26-Like Family C, Member 1A, Cancer/Testis Antigen Family 104, Member 2, CT104.2, POTE Ankyrin Domain Family Member E, POTE2, POTE-2, POTE2gamma, Prostate, Ovary, Testis-Expressed Protein On Chromosome 2, Protein Expressed In Prostate, Ovary, Testis, And Placenta 2
Gene Symbols POTEe
Host Species Rabbit
Immunogen This antibody was developed against a Recombinant Protein corresponding to amino acids:LKALQLNELTMDDDTAVLVIDNGS
Purification Method Immunogen affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 445582
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.