Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POTEE Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | POTEE |
---|---|
Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
POTEE Polyclonal specifically detects POTEE in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
POTEE | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
445582 | |
This antibody was developed against a Recombinant Protein corresponding to amino acids:LKALQLNELTMDDDTAVLVIDNGS | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Human | |
A26C1, A26C1A, ANKRD26-Like Family C Member 1A, ANKRD26-Like Family C, Member 1A, Cancer/Testis Antigen Family 104, Member 2, CT104.2, POTE Ankyrin Domain Family Member E, POTE2, POTE-2, POTE2gamma, Prostate, Ovary, Testis-Expressed Protein On Chromosome 2, Protein Expressed In Prostate, Ovary, Testis, And Placenta 2 | |
POTEe | |
IgG | |
Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title