Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POTEG Antibody (OTI2G1), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N273540A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273540A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273540A647 Supplier Novus Biologicals Supplier No. NBP273540AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

POTEG Monoclonal specifically detects POTEG in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen POTEG
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2G1
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias A26C2, ANKRD26-like family C, member 2, cancer/testis antigen family 104, member 4, CT104.4, POTE ankyrin domain family member G, POTE ankyrin domain family, member G, POTE14ACTBL1, POTE14alpha, POTE-14ANKRD26-like family C member 2, POTE22, Prostate, ovary, testis-expressed protein on chromosome 14, protein expressed in prostate, ovary, testis, and placenta 14
Gene Symbols POTEG
Host Species Mouse
Immunogen Full length recombinant protein of human POTEG (NP_001005356) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 404785.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.