Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POU2F3 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB165141 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165141 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB165141 Supplier Novus Biologicals Supplier No. NBP281774AF405
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

POU2F3 Polyclonal antibody specifically detects POU2F3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen POU2F3
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias Epoc-1, MGC126698, oct-11, OCT11FLJ40063, Octamer-binding protein 11, Octamer-binding transcription factor 11, OTF11, OTF-11, PLA1, PLA-1, POU class 2 homeobox 3, POU domain class 2, transcription factor 3, POU domain, class 2, transcription factor 3, POU transcription factor, Skn-1a, Transcription factor PLA-1, Transcription factor Skn-1
Host Species Rabbit
Immunogen POU2F3 antibody was raised against a 17 amino acid peptide near the carboxy terminus of human POU2F3. The immunogen is located within amino acids 370 - 420 of POU2F3. Amino Acid Squence: SPGNNSRPSSPGSGLH
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25833
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.